Learn More
MCR Safety™ BearKat BK2 Safety Glasses with Clear Lens
Catalog No. 19034409 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Lens Coating:
Anti-fog, Impact-resistant
Catalog No. Lens Coating
19-034-409 Anti-fog, Impact-resistant
1 options
Catalog No. 19-034-409 Supplier MCR Safety™ Supplier No. BK220AF
BearKat™ BK2 series safety glasses with clear lens, UV-AF™ anti-fog coating, soft non-slip pink temple material.
  • Exclusive Duramass™ scratch resistant coating
  • Lightweight frames (20.9 grams) are comfortable and easy to wear
  • Non-slip soft pink temple material
  • Rugged polycarbonate lens filters 99.9% of harmful U.V. rays
  • Single lens design with integral side shields for unobstructed view
  • Small frame
  • Strategically designed for people with smaller profiles
  • UV-AF™ anti-fog coating
  • Meets CAN/CSA Z94.3 standards
  • Meets ANSI/ISEA Z87.1 2020 standard
Specifications
Lens Coating Anti-fog, Impact-resistant
Lens Tint Clear
Product Type Glasses
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection U6
Frame Type Frameless
Autoclavable Not Autoclavable