Learn More
MCR Safety BearKat™ Safety Glasses
Catalog No. 19812729 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Lens Coating:
Impact-resistant, Scratch-resistant
Catalog No. Lens Coating
19-812-729 Impact-resistant, Scratch-resistant
1 options
This item is not returnable. View return policy
Catalog No. 19-812-729 Supplier MCR Safety Supplier No. BK113
This item is not returnable. View return policy
MCR Safety™ BearKat™ Safety Glasses feature a single lens design with integral sideshields for an unobstructed view
  • Light blue single lens
  • Universal nose bridge
  • Reduces glare from artificial light such as halogen and fluorescent.
  • Strong, lightweight design
  • Non-slip soft temple material
  • Exclusive Duramass scratch resistanct coating
  • Rugged Polycarbonate lens filters 99.9% of harmful UV rays
Specifications
Lens Coating Impact-resistant, Scratch-resistant
Lens Tint Blue
Temple Style Hybrid temple with non slip grips
Lens Material Polycarbonate