Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beaucage reagent | 66304-01-6 | 98.0% | 200.24 | C7H4O3S2 | 5 G
SDP

Catalog No. 5000212331
Encompass_Preferred

Beaucage reagent is a small-molecule DNA-cleaving reagent used in nucleic acid chemistry, including conversion of phosphite intermediates to phosphorothioates during oligonucleotide synthesis. It is supplied as a solid or DMSO solution for laboratory research applications.

  • Potent DNA cleavage reagent suitable for nucleic acid chemistry.
  • 98.0% purity by HPLC.
  • Molecular formula C7H4O3S2, molecular weight 200.24.
  • Available as solid and DMSO solution in multiple package sizes.
  • Recommended storage: powder at -20°C for long-term; in solvent at -80°C.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-12331 Supplier Medchemexpress LLC Supplier No. HY1009515G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Beaucage reagent is a small-molecule DNA-cleaving reagent used in nucleic acid chemistry, including conversion of phosphite intermediates to phosphorothioates during oligonucleotide synthesis. It is supplied as a solid or DMSO solution for laboratory research applications.

  • Potent DNA cleavage reagent suitable for nucleic acid chemistry.
  • 98.0% purity by HPLC.
  • Molecular formula C7H4O3S2, molecular weight 200.24.
  • Available as solid and DMSO solution in multiple package sizes.
  • Recommended storage: powder at -20°C for long-term; in solvent at -80°C.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.