Learn More
Cayman Chemical Beauvera basna Basn 100ug
SDP
Catalog No. 502946198
Encompass_Preferred
A fungal metabolite; inhibits Mg2+-, Ca2+-, and Na+/K+-ATPase activities in equine erythrocyte ghosts by 81, 58, and 23%, respectively, at 200 UG/ml; cytotoxic to Sf9 and Sf21 insect cells (CC50s = 4.91 and 12.12 µM, respectively)
Catalog No. 50-294-6198 Supplier Cayman Chemical Supplier No. 32926100 UG
A fungal metabolite; inhibits Mg2+-, Ca2+-, and Na+/K+-ATPase activities in equine erythrocyte ghosts by 81, 58, and 23%, respectively, at 200 UG/ml; cytotoxic to Sf9 and Sf21 insect cells (CC50s = 4.91 and 12.12 µM, respectively)