Learn More
Cayman Chemical Beauvericin, Purity: ≥85%, CAS: 26048-05-5, 10 mg
SDP
Catalog No. 502951894
Encompass_Preferred
A mycotoxin which is cytotoxic against insect cells, killing the insect cell line SF-9 with a 50% cytotoxic concentration of 2.5 μM; induces apoptosis in mammalian cells with an IC50 value of 4.5 μM
Catalog No. 50-295-1894 Supplier Cayman Chemical Supplier No. 1142610 MG
A mycotoxin which is cytotoxic against insect cells, killing the insect cell line SF-9 with a 50% cytotoxic concentration of 2.5 μM; induces apoptosis in mammalian cells with an IC50 value of 4.5 μM