Learn More
AdipoGen Beauvericin
SDP
Supplier:  AdipoGen AGCN20043M001
Encompass_Preferred
Chemical. CAS 26048-05-5. Formula C45H57N3O9. MW 784. Isolated from fungus Beauveria sp. Antibiotic. Apoptosis inducer. Acyl-CoAcholesterol acyltransferase ACAT inhibitor. Anticancer compound. Antihaptotactic and antimetastatic. Antiangiogenic compound. Antibacterial, antiprotozal, antiviral and antifungal compound. Shows ionophoric properties. Cytotoxic. Genotoxic. Potently interacts with ABCB1 and ABCG2 transport functions. Causes mitochondrial dysfunction.
Catalog No. 50-256-2689