Learn More
AdipoGen Beauvericin (5 mg)
SDP
Supplier:  AdipoGen AGCN20043M005
Encompass_Preferred
Chemical. CAS: 26048-05-5. Formula: C45H57N3O9. MW: 784. Isolated from fungus Beauveria sp. Antibiotic. Apoptosis inducer. Acyl-CoA:cholesterol acyltransferase (ACAT) inhibitor. Anticancer compound. Antihaptotactic and antimetastatic. Antiangiogenic compound. Antibacterial, antiprotozal, antiviral and antifungal compound. Shows ionophoric properties. Cytotoxic. Genotoxic. Potently interacts with ABCB1 and ABCG2 transport functions. Causes mitochondrial dysfunction.
Catalog No. 50-205-3391