Learn More
Cayman Chemical BeauverIolIde I 250ug
SDP
Catalog No. 502991172
Encompass_Preferred
A cyclodepsipeptide inhibitor of lipid droplet formation; inhibits lipid droplet formation at 3 and 10 µM and inhibits cholesterol synthesis (IC50 = 0.78 µM) in primary mouse peritoneal macrophages; inhibits ACAT activity in mouse macrophage membranes (IC50 = 6 µM)
Catalog No. 50-299-1172 Supplier Cayman Chemical Supplier No. 31774250 UG
A cyclodepsipeptide inhibitor of lipid droplet formation; inhibits lipid droplet formation at 3 and 10 µM and inhibits cholesterol synthesis (IC50 = 0.78 µM) in primary mouse peritoneal macrophages; inhibits ACAT activity in mouse macrophage membranes (IC50 = 6 µM)