Learn More
Lonza Walkersville BEBM Bronchial Epithelial Cell Growth Basal Medium
Catalog No. NC9556074
Encompass
BEBM is a basal medium sold separately. This is used as a component in the BEGM Bronchial Epithelial Cell Growth Medium BulletKit (catalog number CC-3170) which consists of basal medium and supplements packaged together.BEGM Bronchial Epithelial Cell Growth Mediumis specially designed to support the growth of human primary bronchial epithelial cells. BEGM BulletKit is serum-free and has been optimized to grow cells from normal and diseased tissues.Published research shows BEGM BulletKit has expanded applications. Our customers have used BEGM BulletKit to culture well-established cell lines such as BEAS-2B, home-grown bronchial epithelial cells and supporting co-culture models with breast cancer cell lines such as MCF-7 and MDA-MB-231. Content:1 x Bottle Basal Medium 500 mL
Catalog No. NC9556074 Supplier Lonza Walkersville Supplier No. CC3171
BEBM is a basal medium sold separately. This is used as a component in the BEGM Bronchial Epithelial Cell Growth Medium BulletKit (catalog number CC-3170) which consists of basal medium and supplements packaged together.BEGM Bronchial Epithelial Cell Growth Mediumis specially designed to support the growth of human primary bronchial epithelial cells. BEGM BulletKit is serum-free and has been optimized to grow cells from normal and diseased tissues.Published research shows BEGM BulletKit has expanded applications. Our customers have used BEGM BulletKit to culture well-established cell lines such as BEAS-2B, home-grown bronchial epithelial cells and supporting co-culture models with breast cancer cell lines such as MCF-7 and MDA-MB-231. Content:1 x Bottle Basal Medium 500 mL