Learn More
Medchemexpress LLC HY-19763 5mg Medchemexpress, BEBT-908 CAS:1235449-52-1 Purity:>98%
SDP
Catalog No. 502082180
Encompass_Preferred
Medchemexpress, HY-19763 5mg BEBT-908 CAS:1235449-52-1 BEBT-908 (PI3Kα inhibitor 1) is a selective PI3Kα inhibitor extracted from patent US/20120088764A1, Compound 243, has an IC50<0.1 μM, PI3Kα inhibitor 1 also inhibits HDAC (0.1 μM≤IC50≤1 μM) . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2180 Supplier Medchemexpress LLC Supplier No. HY197635MG
Medchemexpress, HY-19763 5mg BEBT-908 CAS:1235449-52-1 BEBT-908 (PI3Kα inhibitor 1) is a selective PI3Kα inhibitor extracted from patent US/20120088764A1, Compound 243, has an IC50<0.1 μM, PI3Kα inhibitor 1 also inhibits HDAC (0.1 μM≤IC50≤1 μM) . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.