Learn More
Enzo Life Sciences BEC . ammonium salt (1mg). CAS: 63107-40-4 (non-salt)
SDP
Supplier:  Enzo Life Sciences ALX270345M001
Encompass_Preferred
Slow-binding competitive inhibitor of arginase I (Ki=0.4-0.6µM & KD=2.22µM for recombinant rat liver arginase) and arginase II (Ki=0.31µM at pH 7.5 and 0.03µM at pH 9.5 for human recombinant type II arginase). Enhances NO-dependent smooth muscle relaxation in human penile corpus cavernosum tissue. Does not inhibit nitric oxide synthase (NOS) and may serve as a valuable reagent to probe the physiological relationship between arginase and NOS. Inhibitor of rat aortic smooth muscle cell proliferation. Alternative name: S-(2-Boronoethyl)-L-cysteine . NH4. Formula: C5H11BNO4S . NH4. MW: 192.0 . 18.0. Purity: ≥95% (TLC). Appearance: White to off-white powder. Solubility: Soluble in water (10mg/ml). Long Term Storage: +4°C. Handling: Very hygroscopic. Keep under inert gas. Keep cool and dry.
Catalog No. 50-200-8492