Learn More
Abcam BEC, Arginase inhibitor, 1MG
Catalog No. 502705105
Encompass_Preferred
MW 193.03 Da, Purity >98%. Potent, slow binding, competitive arginase inhibitor (Ki values are30 and 500 nM for human arginase II and rat arginase I respectively). Enhances NO-dependent smooth muscle relaxation. Shows pro-inflammatory effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5105 Supplier Abcam Supplier No. AB1456201MG
MW 193.03 Da, Purity >98%. Potent, slow binding, competitive arginase inhibitor (Ki values are30 and 500 nM for human arginase II and rat arginase I respectively). Enhances NO-dependent smooth muscle relaxation. Shows pro-inflammatory effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement