Learn More
Lonza Walkersville HEPES Buffered Saline Solution, 500 mL
Catalog No. NC9616673
Encompass
HEPES Buffered Saline Solution (BSS), is used to neutralize the complex proteins in growth medium that may inactivate trypsin during trypsinization.HEPES BSS at a fill volume of 100 mL is a component of the ReagentPack Subculture kit (catalog number CC-5024). The 100 mL size is also sold separately (catalog number CC-5022).Content:1 x Bottle HEPES Buffered Solution, 500 mL
Catalog No. NC9616673 Supplier Lonza Walkersville Supplier No. CC5024
HEPES Buffered Saline Solution (BSS), is used to neutralize the complex proteins in growth medium that may inactivate trypsin during trypsinization.HEPES BSS at a fill volume of 100 mL is a component of the ReagentPack Subculture kit (catalog number CC-5024). The 100 mL size is also sold separately (catalog number CC-5022).Content:1 x Bottle HEPES Buffered Solution, 500 mL