Learn More
Selleck Chemical LLC BEC HCl
SDP
Catalog No. 5000003607
Encompass_Preferred
BEC HCl is a slow-binding and competitive arginase inhibitor with Ki of 0 31 M (pH7 5) and 0 4-0 6 M for Arginase II and rat Arginase I respectively
Catalog No. 50-000-03607 Supplier Selleck Chemical LLC Supplier No. S792925MG
Add to Cart
Add to Cart
BEC HCl is a slow-binding and competitive arginase inhibitor with Ki of 0 31 M (pH7 5) and 0 4-0 6 M for Arginase II and rat Arginase I respectively