Learn More
AdipoGen Becatecarin
SDP
Supplier:  AdipoGen BVT0258C250
Encompass_Preferred
Chemical. CAS 119673-08-4. Formula C33H34Cl2N4O7. MW 669.6. Semisynthetic. Antibiotic. Semisynthetic water-soluble derivative of rebeccamycin Prod. No. BVT-0139. Antitumor compound. Topoisomerase II Topo II inhibitor. DNA intercalating agent.
Catalog No. 50-256-2033