Learn More
Cayman Chemical BEC (hydrochloride), Purity: ≥95%, CAS: 222638-67-7, 5 mg
SDP
Catalog No. 502987755
Encompass_Preferred
An inhibitor of arginase types I and II; inhibits rat liver arginase I (Kis = 0.4 and 0.6 µM from kinetic analyses); inhibits human arginase II (Kis = 310 and 30 nM at pH 7.5 and 9.5, respectively); enhances nitric oxide-dependent relaxation of human penile corpus cavernosum smooth muscle in vitro at 0.1-1 mM
Catalog No. 50-298-7755 Supplier Cayman Chemical Supplier No. 221455 MG
An inhibitor of arginase types I and II; inhibits rat liver arginase I (Kis = 0.4 and 0.6 µM from kinetic analyses); inhibits human arginase II (Kis = 310 and 30 nM at pH 7.5 and 9.5, respectively); enhances nitric oxide-dependent relaxation of human penile corpus cavernosum smooth muscle in vitro at 0.1-1 mM