Learn More
Beckman Coulter 400 mL Bottle
Catalog No. NC0913146
Encompass
This 400-mL polypropylene bottle is to be used in the SX4400 Swinging Bucket Rotor (part # B01425) for the Allegra X-30 centrifuges.
Catalog No. NC0913146 Supplier Beckman Coulter Supplier No. B01435
This 400-mL polypropylene bottle is to be used in the SX4400 Swinging Bucket Rotor (part # B01425) for the Allegra X-30 centrifuges.