Learn More
Beckman Coulter Beckman Coulter Adapter, Conical Tube, 12 x 15 mL (for Allegra V-15R Swing-Bucket Rotor)
Catalog No. NC3846363
Encompass
Beckman Coulter conical tube adapter designed to hold twelve 15 mL conical tubes per adapter. Supplied as a set of two adapters to enable use of 15 mL tubes in swing-bucket rotor buckets such as the VS 4.750 on Allegra series centrifuges.
  • Holds 12 x 15 mL conical tubes.
  • Supplied as a set of two adapters.
  • Round bucket adapter design for swing-bucket rotors.
  • Compatible with VS 4.750 swing-bucket rotor (Allegra V-15R platform).
  • Allows use of 15 mL tubes for batch processing.
Catalog No. NC3846363 Supplier Beckman Coulter Supplier No. C63164
Beckman Coulter conical tube adapter designed to hold twelve 15 mL conical tubes per adapter. Supplied as a set of two adapters to enable use of 15 mL tubes in swing-bucket rotor buckets such as the VS 4.750 on Allegra series centrifuges.
  • Holds 12 x 15 mL conical tubes.
  • Supplied as a set of two adapters.
  • Round bucket adapter design for swing-bucket rotors.
  • Compatible with VS 4.750 swing-bucket rotor (Allegra V-15R platform).
  • Allows use of 15 mL tubes for batch processing.