Learn More
Beckman Coulter TOC Analyzer, Single Stream, RFID, Quick-Connect (US/Japan power cord)
Catalog No. NC3874752
Encompass
PAT700 is an online total organic carbon (TOC) analyzer designed for continuous monitoring of purified water and water for injection (WFI) applications. It combines TOC, conductivity, resistivity, and temperature measurements in a single instrument and supports regulatory compliance and CIP integration.Single stream configuration for targeted online monitoring.RFID-enabled standards and sample tracking to reduce manual entry.Quick-connect plumbing option for simplified installation.Dual UV lamp with UV detect for reagent-free oxidation and lamp redundancy.Onboard automated standards introduction system for calibration and suitability testing.Dedicated clean-in-place mode for seamless CIP integration.Color touchscreen interface with digital and analog communications.Low consumables and 12-month service interval to reduce operating costs.
Catalog No. NC3874752 Supplier Beckman Coulter Supplier No. D25545
Add to Cart
Add to Cart
Request a Quote
PAT700 is an online total organic carbon (TOC) analyzer designed for continuous monitoring of purified water and water for injection (WFI) applications. It combines TOC, conductivity, resistivity, and temperature measurements in a single instrument and supports regulatory compliance and CIP integration.Single stream configuration for targeted online monitoring.RFID-enabled standards and sample tracking to reduce manual entry.Quick-connect plumbing option for simplified installation.Dual UV lamp with UV detect for reagent-free oxidation and lamp redundancy.Onboard automated standards introduction system for calibration and suitability testing.Dedicated clean-in-place mode for seamless CIP integration.Color touchscreen interface with digital and analog communications.Low consumables and 12-month service interval to reduce operating costs.