Learn More
Beckman Coulter Beckman Coulter Biomek i7 Dual or Hybrid OQ (C30266)
Catalog No. NC3866491
Encompass
Beckman Coulter Biomek i7 Dual or Hybrid OQ (C30266) is an operational qualification (OQ) service intended to verify that Biomek i7 workstations configured with dual-head or hybrid options operate within required performance parameters. The service is designed to provide documented evidence of system operation and functional performance.
  • Operational qualification testing to specified performance criteria.
  • Verification of pipetting accuracy and precision across configured channels.
  • Testing executed on dual-head or hybrid build configurations.
  • Formal documentation of test procedures and results.
Catalog No. NC3866491 Supplier Beckman Coulter Supplier No. C30266
Add to Cart
Add to Cart
Request a Quote
Beckman Coulter Biomek i7 Dual or Hybrid OQ (C30266) is an operational qualification (OQ) service intended to verify that Biomek i7 workstations configured with dual-head or hybrid options operate within required performance parameters. The service is designed to provide documented evidence of system operation and functional performance.
  • Operational qualification testing to specified performance criteria.
  • Verification of pipetting accuracy and precision across configured channels.
  • Testing executed on dual-head or hybrid build configurations.
  • Formal documentation of test procedures and results.