Learn More
Beckman Coulter Beckman Coulter FX6100 Fixed-Angle Aluminum Rotor - 6 x 100 mL (for Allegra X-15R / X-12 centrifuges)
Catalog No. NC3902659
Encompass
The FX6100 fixed-angle aluminum rotor is a bio-certified rotor designed for density gradient and differential separations. It accepts six 100 mL tubes and is rated for high-speed centrifugation, suitable for separations of erythrocytes, cell lysate fractions, and biomolecule isolation.
  • Accommodates six 100 mL tubes with a nominal 600 mL capacity.
  • Bio-certified for safe handling of biological samples.
  • Aluminum construction for reliable performance and reduced corrosion.
  • Compatible with Allegra X-15R and Allegra X-12 series centrifuges.
  • Rated to 10,200 rpm (11,400 x g) for high-speed separations.
  • Accepts tubes 38 x 102 mm.
Catalog No. NC3902659 Supplier Beckman Coulter Supplier No. 369735
The FX6100 fixed-angle aluminum rotor is a bio-certified rotor designed for density gradient and differential separations. It accepts six 100 mL tubes and is rated for high-speed centrifugation, suitable for separations of erythrocytes, cell lysate fractions, and biomolecule isolation.
  • Accommodates six 100 mL tubes with a nominal 600 mL capacity.
  • Bio-certified for safe handling of biological samples.
  • Aluminum construction for reliable performance and reduced corrosion.
  • Compatible with Allegra X-15R and Allegra X-12 series centrifuges.
  • Rated to 10,200 rpm (11,400 x g) for high-speed separations.
  • Accepts tubes 38 x 102 mm.