Learn More
HemoCue America Beckman Coulter™ Gastroccult™ Gastric Sample Occult Blood Test Kit
GSA_VA
Catalog No. SK66040
Change view
Click to view available options
Quantity:
40 Pack
40 Tests
6 x 15 mL
Type:
Gastroccult Developer
Occult Blood Test Kit
Straw Applicator
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
SK-66040 40 Tests Occult Blood Test Kit
SK-66115 6 x 15 mL Gastroccult Developer
SK-66140 40 Pack Straw Applicator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. SK-66040 Supplier HemoCue America Supplier No. 66040A
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Two tests in one
  • CLIA Complexity: Waived
  • CPT Codes: 87723 for occult blood, 83986 for pH
  • Slide test for detecting occult blood and determining pH in gastric samples, both aspirate and vomitus, on the same test card
  • On-slide Performance Monitor assures that the occult blood test is performing to product specifications
  • Scale of pH test ranges from a clinically useful 1.3 to 7.8
  • Both test results are available within one minute
  • Test cards, developer, and applicator straws sold separately
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
CPT Code 87723
Clia Complexity Waived
Detectable Analytes Occult Blood
Type Occult Blood Test Kit
Includes Convenient pH comparison chart, on-slide performance Monitor™, 40 Gastroccult™ Tests and instructions
Quantity 40 Tests
Test Time 1 min.