Learn More
HemoCue America Beckman Coulter™ Hemoccult™ ICT 2-Day Patient Screening Kit
Catalog No. 23280035
Change view
Click to view available options
Quantity:
50 Pack
Catalog No. Quantity
23-280-035 50 Pack
1 options
This item is not returnable. View return policy
Catalog No. 23-280-035 Supplier HemoCue America Supplier No. 395261A
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Dispensing Envelope with patient instructions; Hemoccult ICT Collection Cards; Flushable Collection Tissues; Applicator Sticks; Mailing Pouches
Immunochemical FOBT (iFOBT) with high specificity and sensitivity
  • Clinically proven to detect bleeding associated with more cancers and polyps than traditional guaiac-based FOBTs
  • An ideal screening tool for colorectal cancer due to superior specificity for lower GI bleeding
  • Human hemoglobin specificity reduces the number of false positive test results
  • No diet or drug restrictions
  • Safe, non-invasive
  • CLIA waived with clear, readable results
TRUSTED_SUSTAINABILITY
Specifications
CPT Code G0328QW, 82274QW
Clia Complexity Waived
Type ICT 2-day Patient Screening Kit
For Use With (Application) Detect bleeding associated with cancers and polyps
Includes Dispensing envelopes with instructions, collection cards, collection tissues, applicator sticks, mailing pouches
Quantity 50 Pack