Learn More
HemoCue America Beckman Coulter™ Hemoccult™ ICT Immunochemical Fecal Occult Blood Test Kit
GSA_VA
Catalog No. 23280010
Change view
Click to view available options
Quantity:
100 Pack
40 Pack
Type:
ICT Collection Cards
ICT Sample Collection Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23280010 100 Pack ICT Collection Cards
23280011 40 Pack ICT Sample Collection Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23280010 Supplier HemoCue America Supplier No. 395065A
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
ICT FOBT Kit (Cat. No. 23-280-012) includes test devices and 8mL bottle of Hemoccult ICT Buffer; ICT Sample Collection Kit (Cat. No. 23-280-011) contains Patient Kits; each Patient Kit contains Dispensing Envelope with Patient Instructions, Hemoccult ICT Collection Cards, Collection Tissues, Applicator Sticks, and Mailing Pouch; ICT Collection Cards (Cat. No. 23-280-010) contain Collection Cards and Applicator Sticks; ICT Control Kit (Cat. No. 23-280-013) contains bottles (Positive and Negative)
  • CLIA Complexity: Waived
  • CPT Codes: 82274QW, G0328QW
  • Rapid, qualitative immunochemical chromatographic method for detection of human hemoglobin from blood in fecal samples
  • Visually read results
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
CPT Code 82274QW, G0328QW
Clia Complexity Waived
Type ICT Collection Cards
For Use With (Application) Detection of Human hemoglobin from Blood in fecal samples
Includes Collection cards and applicator sticks
Quantity 100 Pack