Learn More
HemoCue America Beckman Coulter™ Hemoccult™ SENSA™ Fecal Occult Blood Slide Test System
Catalog No. SK64152
Change view
Click to view available options
Quantity:
100 Pack
1000 Case
40 Pack
Type:
Dispensapak Plus Kit
Single Slide Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
SK-64152 1000 Case Single Slide Kit
SK-64130 40 Pack Dispensapak Plus Kit
SK-64151 100 Pack Single Slide Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. SK-64152 Supplier HemoCue America Supplier No. 64152
Add to Cart
Add to Cart
This item is not returnable. View return policy
Provides enhanced detection at low, but abnormal, levels of hemoglobin
  • CLIA Complexity: Waived
  • CPT Codes: 82270, G0107
  • Convenient and qualitative method for detecting fecal occult blood, which may be indicative of gastrointestinal disease such as colon cancer
  • Vivid blue reaction provides a high level of color stability and intensity, resulting in superior readability for greater accuracy and ease of interpretation
  • Recommended for professional use as a diagnostic aid during routine physical examinations, for hospital patients to monitor for bleeding in patients with iron deficiency anemia or recuperating from surgery, peptic ulcer, ulcerative colitis and other conditions
  • The Hemoccult II SENSA Dispensapak Plus is ideal for screening programs for colorectal cancer when the patient instructions are closely followed
  • Hemoccult II Slides feature cards of three tests designed to allow patients to collect three serial specimens at home
Order Info
Supplies and Developer Kit available separately (Cat. Nos. 23-280-241 and SK64115).
TRUSTED_SUSTAINABILITY
Specifications
CPT Code 82270, G0107
Clia Complexity Waived
Type Single Slide Kit
For Use With (Application) Detection of fecal occult Blood
Includes 100 SENSA single slides, 20 x 15mL SENSA developer and 1000 applicator sticks
Quantity 1000 Case