Learn More
Beckman Coulter™ HemosIL™ Reference Emulsion
Catalog No. IL9756904
Change view
Click to view available options
Quantity:
4 x 400 mL
Catalog No. Quantity
IL9756904 4 x 400 mL
1 options
This item is not returnable. View return policy
Catalog No. IL9756904 Supplier Beckman Coulter™ Supplier No. 9756904
This item is not returnable. View return policy
For nephelometric analysis and as a washing solution on the ACL 100-7000 Series Coagulation Analyzers
Specifications
Type Beckman Coulter HemosIL Reference Emulsion
For Use With (Equipment) ACL Coagulation Analyzers
Capacity (Metric) 4 x 400 mL
Quantity 4 x 400 mL