Learn More
HemoCue America Beckman Coulter ICON™ II HCG Combo Latex Test Kit for Serum and Urine
Catalog No. 23280000
Change view
Click to view available options
Test Kit
Catalog No.
23-280000 Test Kit
1 options
This item is not returnable. View return policy
Catalog No. 23-280000 Supplier HemoCue America Supplier No. 4135
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Dual-test capability eliminates the need to stock separate kits
  • CPT Codes: 81025, 84702, 84703
  • CLIA Complexity: Waived for urine
  • Semiquantitative for serum hCG; qualitative for urine hCG
  • hCG filter membrane has a 25mIU/mL calibrated internal reference zone, a patient test zone with a detection sensitivity of 10mIU/mL for serum (20mIU/mL for urine) and a negative control zone that indicates interfering substances (invalid test)
Order Info
Controls are packaged two vials per level (positive and negative. Supplied in package with sufficient material to perform 50 tests