Learn More
HemoCue America Beckman Coulter ICON™ II HCG for Urine
Catalog No. 23272744
Change view
Click to view available options
Quantity:
6 x 3 mL
Catalog No. Quantity
23-272744 6 x 3 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-272744 Supplier HemoCue America Supplier No. 9115
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Semiquantitative results with a 50mIU/mL reference and a test sensitivity of 20mIU/mL
  • Semiquantitative for urine hCG
  • CLIA Complexity: Waived
  • Precise and easy-to-read results
  • Patient test zone located in center of membrane and internal reference zone located at the “12 o'clock” position
  • Blue dot of any intensity in the patient test zone indicates a positive result
  • Color comparison on positive blue dot with internal reference provides semiquantitative test results
  • Absence of a circular blue dot in the patient test zone indicates negative result
Order Info
Supplied in package with sufficient material for 24 or 50 tests. Controls are packaged three vials per level (positive and negative)
TRUSTED_SUSTAINABILITY
Specifications
Quantity 6 x 3 mL