Learn More
HemoCue America Beckman Coulter™ ICON™ Mono Test Kit
GSA_VA
Catalog No. 23280016
Change view
Click to view available options
Quantity:
30 Tests
Catalog No. Quantity
23-280-016 30 Tests
1 options
This item is not returnable. View return policy
Catalog No. 23-280-016 Supplier HemoCue America Supplier No. 395100A
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
  • 30 Tests
  • 30 Capillary Tubes
  • 1 Developer Solution
  • 1 Each Positive and Negative Control
  • 1 Product Instruction
A one-step antibody test for fast, accurate and easy-to-read result at the-point-of care
For use on patients who present with fever, sore throat, swollen lymph glands or other symptoms common with a viral infection and may not be able to distinguish these from infectious mononucleosis (IM).
  • Approved for testing on all ages
  • Combines finger-stick whole blood convenience with EIA membrane technology, for fast, accurate results
  • Uses direct solid-phase immunoassay technology for early qualitative detection of IM heterophile antibodies in whole blood, serum or plasma
  • 99.9% relative sensitivity and 98.8% relative specificity serum/plasma; 98.3% relative specificity WB assay
  • CLIA Category
    • Whole Blood - Waived
    • Serum/Plasma - Moderately Complex
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Clia Complexity Waived
Detectable Analytes Heterophile IgM
Type Mononucleosis Test Kit
Includes 30 Tests, 30 capillary tubes, 1 developer solution, 1 each positive and negative control, 1 product instruction
Quantity 30 Tests