Learn More
Beckman Coulter Beckman Coulter JS-5.3 Swinging-Bucket Rotor with Microplate Kit for Avanti J-E and J-26 XP centrifuges
Catalog No. NC3854726
Encompass_Preferred
The JS-5.3 swinging-bucket aluminum rotor package is a four-place, high-performance rotor designed for Avanti-series centrifuge platforms. It is supplied with buckets and a microplate carrier kit and accepts adapters to accommodate a wide range of tubes, bottles, and microplates. The rotor is rated to 5,300 rpm and provides reliable RCF for preparative applications.
  • High-performance swinging-bucket design.
  • Anodized aluminum construction for durability.
  • Rated to 5,300 rpm; up to 6,870 x g depending on configuration.
  • Four-place capacity with adapters for tubes, bottles, and microplates.
  • Includes buckets, tie-down knob, and microplate carrier kit.
Catalog No. NC3854726 Supplier Beckman Coulter Supplier No. 969314
Add to Cart
Add to Cart
Request a Quote
The JS-5.3 swinging-bucket aluminum rotor package is a four-place, high-performance rotor designed for Avanti-series centrifuge platforms. It is supplied with buckets and a microplate carrier kit and accepts adapters to accommodate a wide range of tubes, bottles, and microplates. The rotor is rated to 5,300 rpm and provides reliable RCF for preparative applications.
  • High-performance swinging-bucket design.
  • Anodized aluminum construction for durability.
  • Rated to 5,300 rpm; up to 6,870 x g depending on configuration.
  • Four-place capacity with adapters for tubes, bottles, and microplates.
  • Includes buckets, tie-down knob, and microplate carrier kit.