Learn More
Beckman Coulter Beckman Coulter lid O-ring, Buna-N, 7.737 in ID (for JA-14.50 rotor lid)
Catalog No. NC3806360
Encompass
Replacement large lid O-ring for Beckman Coulter centrifuge rotors. Supplied in Buna-N (nitrile) rubber and sized to the AS568-170 standard to provide a resilient, oil-resistant seal for rotor lids.
  • Made from Buna-N (nitrile) rubber.
  • Inner diameter approximately 7.737 in (196.5 mm).
  • Conforms to AS568-170 standard size.
  • Resists petroleum-based oils and greases.
  • Provides a resilient, reusable seal for rotor lids.
Catalog No. NC3806360 Supplier Beckman Coulter Supplier No. 885501
Replacement large lid O-ring for Beckman Coulter centrifuge rotors. Supplied in Buna-N (nitrile) rubber and sized to the AS568-170 standard to provide a resilient, oil-resistant seal for rotor lids.
  • Made from Buna-N (nitrile) rubber.
  • Inner diameter approximately 7.737 in (196.5 mm).
  • Conforms to AS568-170 standard size.
  • Resists petroleum-based oils and greases.
  • Provides a resilient, reusable seal for rotor lids.