Learn More
Fluotics iSeries/FX/NX Compatible P80, sterile
Catalog No. 5000435255
Encompass
Beckman iSeries/FX/NX Compatible P80, sterile, Working Volume: 80uL, Tip Color: Clear, Tip Box, 96 Multi Channel Head, Compare To: 717255, B85767, 50 racks/case with 96 tips per rack
Catalog No. 50-004-35255 Supplier Fluotics Supplier No. BFXI80.ST
Beckman iSeries/FX/NX Compatible P80, sterile, Working Volume: 80uL, Tip Color: Clear, Tip Box, 96 Multi Channel Head, Compare To: 717255, B85767, 50 racks/case with 96 tips per rack