Learn More
Medchemexpress LLC HY-13571A 250mg Medchemexpress, Beclometasone dipropionate CAS:5534-09-8 Purity:>98%
SDP
Catalog No. 502081399
Encompass_Preferred
Medchemexpress, HY-13571A 250mg Beclometasone dipropionate CAS:5534-09-8 Betamethasone dipropionate, the prodrug of Betamethasone, is an orally active and potent glucocorticoid with anti-inflammatory and immunosuppressive activity. Betamethasone appears to be an effective inhibitor of LPS-induced inflammation and MMP release. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1399 Supplier Medchemexpress LLC Supplier No. HY13571A250MG
Medchemexpress, HY-13571A 250mg Beclometasone dipropionate CAS:5534-09-8 Betamethasone dipropionate, the prodrug of Betamethasone, is an orally active and potent glucocorticoid with anti-inflammatory and immunosuppressive activity. Betamethasone appears to be an effective inhibitor of LPS-induced inflammation and MMP release. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.