Learn More
Cayman Chemical Beclomethasone, Purity: ≥95%, CAS: 4419-39-0, 100 mg
SDP
Catalog No. 502949538
Encompass_Preferred
A glucocorticoid; reduces plasma corticosteroid levels in rats from 1-10 mg/kg
Catalog No. 50-294-9538 Supplier Cayman Chemical Supplier No. 33196100 MG
A glucocorticoid; reduces plasma corticosteroid levels in rats from 1-10 mg/kg