Learn More
Beclomethasone dipropionate, ≥99%, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Chemical Name or Material | Beclomethasone Dipropionate |
| CAS | 9-8-5534 |
| Physical Form | Neat |
| Linear Formula | C28H37 ClO7 |
| Technique | Gas Chromatography |
| MDL Number | MFCD00135613 |
| Synonym | 9-Chloro-11 beta,17,21-trihydroxy-16 beta-methylpregna-1,4-diene-3,20-dione 17,21-dipropionate |
| Type | Analytical Standard |
| Solvent or Matrix | Neat |
| Molecular Weight (g/mol) | 521.04 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.