Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beclomethasone Dipropionate, MP Biomedicals™

Catalog No. ICN15481494 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15481494 5 g
ICN15481491 1 g
ICN15481483 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15481494 Supplier MP Biomedicals, Inc Supplier No. 0215481494
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antiallergic, antiasthmatic, anti-inflammatory
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00151238

Specifications

CAS 1327543
Color White
Molecular Formula C10H12N5Na2O8P
Quantity 5 g
Synonym guanosine 5'-monophosphate disodium salt, 5'-guanylic acid disodium salt, disodium guanylate, disodium 5'-guanylate, sodium guanylate, disodium gmp, disodium 5'-gmp, gmp disodium salt, 5'-guanylic acid, disodium salt, 5'-gmp disodium salt
InChI Key PVBRXXAAPNGWGE-KHRSEZDTNA-L
SMILES [Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]2O)C(=O)N1
IUPAC Name disodium [(2R,3S,4R,5R)-5-(2-amino-6-oxo-6,9-dihydro-1H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
Molecular Weight (g/mol) 407.19
PubChem CID 21712
Formula Weight 521
Physical Form Solid
Chemical Name or Material Beclomethasone Dipropionate
Show More Show Less
storageNote1 Protect from light.
Recommended Storage Store at +4°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.