Learn More
Description
Beclomethasone Dipropionate USP (Micronized) is a white to cream white, odorless powder. Micronized with a particle size distribution of 90% ≤ 5 μm.
Alternative name(s): Beclometasone Dipropionate
Specifications
Specifications
| Chemical Name or Material | Beclomethasone Dipropionate, USP (Micronized) |
| Packaging | Jar 60ml, White, HDPE, Round, 33-400, TRIVENI-DB, CT |
| CAS | 5534-09-8 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 521.04 |
| Quantity | 5 g |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
