Learn More
Aviva Systems Biology Corp BECN1 Antibody - N-terminal region (ARP58595_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58595_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BECN1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human BECN1. Alternate names: ATG6, VPS30, beclin1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62365