Learn More
Selleck Chemical LLC Bedaquiline 10mg 843663-66-1 TMC-207, R207910
SDP
Catalog No. 501948406
Encompass_Preferred
Bedaquiline (TMC-207, R207910), a diarylquinoline, binds to subunit c of mycobacterial ATP synthase and inhibits its activity. It is highly selective for mycobacterial ATP synthase enzyme compared to homologous eukaryotic enzyme (Selectivity Index >20 000). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8406 Supplier Selleck Chemical LLC Supplier No. S562310MG
Bedaquiline (TMC-207, R207910), a diarylquinoline, binds to subunit c of mycobacterial ATP synthase and inhibits its activity. It is highly selective for mycobacterial ATP synthase enzyme compared to homologous eukaryotic enzyme (Selectivity Index >20 000). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.