Learn More
Medchemexpress LLC HY-14881 50mg , Bedaquiline TMC207;R207910 CAS:843663-66-1 Purity:98%
SDP
Catalog No. 501493745
Encompass_Preferred
Medchemexpress, HY-14881 50mg Bedaquiline TMC207;R207910 CAS:843663-66-1 Formula:C32H31BrN2O2 Purity:98% Bedaquiline, a diarylquinoline antibiotic that targets ATP synthase, is effective for the treatment of Mycobacterium tuberculosis infections. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3745 Supplier Medchemexpress LLC Supplier No. HY1488150MG
Medchemexpress, HY-14881 50mg Bedaquiline TMC207;R207910 CAS:843663-66-1 Formula:C32H31BrN2O2 Purity:98% Bedaquiline, a diarylquinoline antibiotic that targets ATP synthase, is effective for the treatment of Mycobacterium tuberculosis infections. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.