Learn More
Apexbio Technology LLC Bedaquiline 843663-66-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062800
Encompass_Preferred
Bedaquiline a diarylquinoline antibiotic targets Mycobacterium tuberculosis via dual inhibition of subunits c and within the bacterial F1FO-ATP synthase complex disrupting ATP production It is applied primarily in research addressing multi-drug resistant (MDR) tuberculosis Recent studies have expanded its use into oncology where bedaquiline inhibits mitochondrial respiratory function and reduces glycolytic activity in cancer stem-cell-like populations In vitro experiments employing breast cancer MCF7 cells revealed bedaquiline increased ROS levels and reduced mitochondrial membrane potential Reported IC50 values for growth inhibition range approximately 1-10 M depending on cell lines and experimental conditions
Catalog No. 50-000-62800 Supplier Apexbio Technology LLC Supplier No. B34925.1
Bedaquiline a diarylquinoline antibiotic targets Mycobacterium tuberculosis via dual inhibition of subunits c and within the bacterial F1FO-ATP synthase complex disrupting ATP production It is applied primarily in research addressing multi-drug resistant (MDR) tuberculosis Recent studies have expanded its use into oncology where bedaquiline inhibits mitochondrial respiratory function and reduces glycolytic activity in cancer stem-cell-like populations In vitro experiments employing breast cancer MCF7 cells revealed bedaquiline increased ROS levels and reduced mitochondrial membrane potential Reported IC50 values for growth inhibition range approximately 1-10 M depending on cell lines and experimental conditions