Learn More
Medchemexpress LLC HY-14881A 50mg , Bedaquiline (fumarate) CAS:845533-86-0 Purity:>98%
SDP
Catalog No. 502259736
Encompass_Preferred
HY-14881A 50mg Bedaquiline (fumarate) CAS: 845533-86-0 Bedaquiline fumarate, a diarylquinoline antibiotic that targets ATP synthase, is effective for the treatment of Mycobacterium tuberculosis infections. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9736 Supplier Medchemexpress LLC Supplier No. HY14881A50MG
HY-14881A 50mg Bedaquiline (fumarate) CAS: 845533-86-0 Bedaquiline fumarate, a diarylquinoline antibiotic that targets ATP synthase, is effective for the treatment of Mycobacterium tuberculosis infections. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.