Learn More
Apexbio Technology LLC Bedaquiline fumarate 845533-86-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000092438
Encompass_Preferred
Bedaquiline fumarate (CAS 845533-86-0) is a small-molecule inhibitor targeting ATP synthase of Mycobacterium tuberculosis It is designed to inhibit mycobacterial ATP synthase thereby disrupting ATP biosynthesis and bacterial energy metabolism Bedaquiline fumarate exerts its biological activity primarily through selective inhibition of ATP synthase In biochemical assays bedaquiline fumarate demonstrates potent inhibitory activity with an IC50 value of approximately 30 nM against M tuberculosis ATP synthase Based on these pharmacological properties bedaquiline fumarate holds research potential in studying tuberculosis pathogenesis mechanisms of antibiotic resistance and evaluating therapeutic strategies for mycobacterial infections
Catalog No. 50-000-92438 Supplier Apexbio Technology LLC Supplier No. B34915.1
Bedaquiline fumarate (CAS 845533-86-0) is a small-molecule inhibitor targeting ATP synthase of Mycobacterium tuberculosis It is designed to inhibit mycobacterial ATP synthase thereby disrupting ATP biosynthesis and bacterial energy metabolism Bedaquiline fumarate exerts its biological activity primarily through selective inhibition of ATP synthase In biochemical assays bedaquiline fumarate demonstrates potent inhibitory activity with an IC50 value of approximately 30 nM against M tuberculosis ATP synthase Based on these pharmacological properties bedaquiline fumarate holds research potential in studying tuberculosis pathogenesis mechanisms of antibiotic resistance and evaluating therapeutic strategies for mycobacterial infections