Learn More
Medchemexpress LLC Bedaquiline (fumarate) | 845533-86-0 | 671.58 | 100 MG
SDP
Catalog No. 5000361504
Encompass_Preferred
Bedaquiline fumarate is an analytical standard and a diarylquinoline antibiotic that targets ATP synthase, effective for treating *Mycobacterium tuberculosis* infections. This product is intended for research and analytical applications.
  • Effective for treating *Mycobacterium tuberculosis* infections.
  • Inhibits growth of TDR *M. tuberculosis* strains.
  • Shows activity against *Mycobacterium avium*.
  • Active against *Mycobacterium abscessus* subspecies *abscessus* and *massiliense*.
  • Possesses moderate in vitro activity against NTM species.
  • Demonstrates excellent in vitro activity against multidrug resistant *M. tuberculosis*.
Catalog No. 50-003-61504 Supplier Medchemexpress LLC Supplier No. HY14881AR100MG
Bedaquiline fumarate is an analytical standard and a diarylquinoline antibiotic that targets ATP synthase, effective for treating *Mycobacterium tuberculosis* infections. This product is intended for research and analytical applications.
  • Effective for treating *Mycobacterium tuberculosis* infections.
  • Inhibits growth of TDR *M. tuberculosis* strains.
  • Shows activity against *Mycobacterium avium*.
  • Active against *Mycobacterium abscessus* subspecies *abscessus* and *massiliense*.
  • Possesses moderate in vitro activity against NTM species.
  • Demonstrates excellent in vitro activity against multidrug resistant *M. tuberculosis*.