Learn More
Medchemexpress LLC Bedaquiline fumarate | 845533-86-0 | 99.98% | 671.58 | 50 MG
SDP
Catalog No. 5000361482
Encompass_Preferred
Bedaquiline fumarate is an analytical standard and a diarylquinoline antibiotic that targets ATP synthase. It is effective for the treatment of Mycobacterium tuberculosis infections and is intended for research and analytical applications.
  • Analytical standard grade
  • Reference standard supplied assay
  • Effective against Mycobacterium tuberculosis infections
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-003-61482 Supplier Medchemexpress LLC Supplier No. HY14881AR50MG
Bedaquiline fumarate is an analytical standard and a diarylquinoline antibiotic that targets ATP synthase. It is effective for the treatment of Mycobacterium tuberculosis infections and is intended for research and analytical applications.
  • Analytical standard grade
  • Reference standard supplied assay
  • Effective against Mycobacterium tuberculosis infections
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications