Learn More
Medchemexpress LLC Bedaquiline (fumarate) | 845533-86-0 | 99.99% | 671.58 | 25 MG
SDP
Catalog No. 5000363448
Encompass_Preferred
Bedaquiline (fumarate) is a diarylquinoline antibiotic that targets ATP synthase and is effective for the treatment of Mycobacterium tuberculosis infections. It inhibits the growth of TDR M. tuberculosis strains and shows high activity against Mycobacterium avium and moderate activity against NTM species. This product is for research use only.
  • Targets ATP synthase
  • Effective for treating Mycobacterium tuberculosis infections
  • Inhibits growth of TDR M. tuberculosis strains
  • High activity against Mycobacterium avium
  • Moderate activity against NTM species
  • For research use only
Catalog No. 50-003-63448 Supplier Medchemexpress LLC Supplier No. 25MGHY14881AR
Bedaquiline (fumarate) is a diarylquinoline antibiotic that targets ATP synthase and is effective for the treatment of Mycobacterium tuberculosis infections. It inhibits the growth of TDR M. tuberculosis strains and shows high activity against Mycobacterium avium and moderate activity against NTM species. This product is for research use only.
  • Targets ATP synthase
  • Effective for treating Mycobacterium tuberculosis infections
  • Inhibits growth of TDR M. tuberculosis strains
  • High activity against Mycobacterium avium
  • Moderate activity against NTM species
  • For research use only