Learn More
Medchemexpress LLC Bedaquinoline impurity 1 | 1972612-60-4 | 555.50 g/mol | C32H31BrN2O2 | 10 MG
SDP
Catalog No. 5000285650
Encompass_Preferred
Bedaquinoline impurity 1 is an identified impurity of the anti-tuberculosis agent bedaquiline, provided as an analytical reference for research and impurity profiling. The compound has molecular formula C32H31BrN2O2 and molecular weight 555.50 g/mol. It is supplied in small milligram quantities for laboratory analytical use.
  • Analytical reference for impurity identification and method development.
  • Useful for chromatographic and mass spectrometric characterization.
  • Supplied in small, defined quantities suitable for lab-scale analysis.
  • Chemical identity documented with CAS number and molecular formula for traceability.
Catalog No. 50-002-85650 Supplier Medchemexpress LLC Supplier No. HY14881E10MG
Bedaquinoline impurity 1 is an identified impurity of the anti-tuberculosis agent bedaquiline, provided as an analytical reference for research and impurity profiling. The compound has molecular formula C32H31BrN2O2 and molecular weight 555.50 g/mol. It is supplied in small milligram quantities for laboratory analytical use.
  • Analytical reference for impurity identification and method development.
  • Useful for chromatographic and mass spectrometric characterization.
  • Supplied in small, defined quantities suitable for lab-scale analysis.
  • Chemical identity documented with CAS number and molecular formula for traceability.