Learn More
Medchemexpress LLC Bedaquinoline impurity 1 | 1972612-60-4 | >98.6% | 555.50 | 50 MG
SDP
Catalog No. 5000361503
Encompass_Preferred
Bedaquinoline impurity 1 is an impurity of Bedaquinoline. It is supplied as a solid for research use only.
  • Supplied as a solid
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month
  • Ships at room temperature in continental US
Catalog No. 50-003-61503 Supplier Medchemexpress LLC Supplier No. HY14881E50MG
Bedaquinoline impurity 1 is an impurity of Bedaquinoline. It is supplied as a solid for research use only.
  • Supplied as a solid
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month
  • Ships at room temperature in continental US