Learn More
Fine Science Tools Beebee Bone Scissors, Stainless Steel, 10 cm
SDP
Catalog No. NC9851541
Encompass
Tip Shape: Straight/ Cutting Edge: 15mm/ Alloy / Material: Stainless Steel/ Length: 10cm/ Joint Action: Single
Catalog No. NC9851541 Supplier Fine Science Tools Supplier No. 1604410
Tip Shape: Straight/ Cutting Edge: 15mm/ Alloy / Material: Stainless Steel/ Length: 10cm/ Joint Action: Single