Learn More
Alkali Scientific Beef Extract, 10 Kg
SDP
Catalog No. 5000662298
Encompass_Preferred
Beef Extract is a microbiological nutrient ingredient used in the preparation of culture media for bacterial and microbial growth. It contains naturally derived peptides, amino acids, minerals, and vitamins that provide essential nutrients for microbial metabolism. When added to laboratory culture media, beef extract supports the development of bacterial colonies and enhances the growth of many non fastidious microorganisms. It is commonly used in general purpose media such as nutrient broth, nutrient agar, and other microbiological formulations. Researchers rely on beef extract to maintain stable and reproducible culture conditions in microbiology laboratories, fermentation studies, and microbial physiology experiments where consistent nutrient availability is required.
Catalog No. 50-006-62298 Supplier Alkali Scientific Supplier No. BEE004PL
Beef Extract is a microbiological nutrient ingredient used in the preparation of culture media for bacterial and microbial growth. It contains naturally derived peptides, amino acids, minerals, and vitamins that provide essential nutrients for microbial metabolism. When added to laboratory culture media, beef extract supports the development of bacterial colonies and enhances the growth of many non fastidious microorganisms. It is commonly used in general purpose media such as nutrient broth, nutrient agar, and other microbiological formulations. Researchers rely on beef extract to maintain stable and reproducible culture conditions in microbiology laboratories, fermentation studies, and microbial physiology experiments where consistent nutrient availability is required.